missing translation for 'onlineSavingsMsg'
Learn More

L-Alanyl-L-glutamine, 99%

L-Alanyl-L-glutamine, CAS # 39537-23-0, is a synthesized dipeptide, which can be digested to form stable L-glutamine in vitro. | CAS: 39537-23-0 | C8H15N3O4 | 217.23 g/mol

$87.74 - $771.44

Chemical Identifiers

CAS 39537-23-0
Molecular Formula C8H15N3O4
Molecular Weight (g/mol) 217.23
MDL Number MFCD00133046
InChI Key HJCMDXDYPOUFDY-WHFBIAKZSA-N
Synonym l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax
PubChem CID 123935
ChEBI CHEBI:73788
SMILES C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6699606
View Documents
Thermo Scientific Chemicals
J6699606
5 g
Each for $87.74
Only null left
 
AAJ6699614
View Documents
Thermo Scientific Chemicals
J6699614
25 g
Each for $250.67
Only null left
 
AAJ6699622
View Documents
Thermo Scientific Chemicals
J6699622
100 g
Each for $771.44
Only null left
 
Description

Description

A substitute for glutamine in mammalian cell culture media; stable to heat-sterilization.L-Alanyl-L-glutamine is used as a component in pharmaceuticals, cosmetics and food products. It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency. It provides nutritional requirements, improves intestinal health and enhances the immune system. It is also involved in various clinical studies such as cancer treatment, radiation and chemotherapy. It finds application in biotechnology as a component for mammalian cell culture medium. It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Alanyl-L-glutamine is a synthetic dipeptide that degrades to L-glutamine

Application

• L-Alanyl-L-glutamine can be used as a substitute for glutamine in mammalian cell culture media and is stable to heat-sterilization
• It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency
• It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release
• When this dipeptide is added to cultures of antibody-producing CHO cells in vitro, it reduces apoptosis and promotes antibody production

Specifications

Chemical Identifiers

39537-23-0
217.23
HJCMDXDYPOUFDY-WHFBIAKZSA-N
123935
C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O
C8H15N3O4
MFCD00133046
l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax
CHEBI:73788

Specifications

222°C
615°C
C8H15N3O4
l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax
HJCMDXDYPOUFDY-WHFBIAKZSA-N
(2S)-2-[(2S)-2-aminopropanamido]-4-carbamoylbutanoic acid
5 g
123935
217.22
Powder
39537-23-0
White
MFCD00133046
Soluble in water.
C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O
217.23
>110°C (230°F)
CHEBI:73788
99%
L-Alanyl-L-glutamine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 475-290-9

RTECSNumber : MA2275262

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only