missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Alanyl-L-glutamine, 97%
CAS: 39537-23-0 | C8H15N3O4 | 217.23 g/mol
$309.05 - $309.05
Chemical Identifiers
| CAS | 39537-23-0 |
|---|---|
| Molecular Formula | C8H15N3O4 |
| Molecular Weight (g/mol) | 217.23 |
| MDL Number | MFCD00133046 |
| InChI Key | HJCMDXDYPOUFDY-WHFBIAKZSA-N |
| Synonym | l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax |
| PubChem CID | 123935 |
| ChEBI | CHEBI:73788 |
| IUPAC Name | (2S)-5-amino-2-[[(2S)-2-aminopropanoyl]amino]-5-oxopentanoic acid |
| SMILES | C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC451610050
|
Thermo Scientific Chemicals
451610050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 39537-23-0 | |
| 217.23 | |
| HJCMDXDYPOUFDY-WHFBIAKZSA-N | |
| 123935 | |
| (2S)-5-amino-2-[[(2S)-2-aminopropanoyl]amino]-5-oxopentanoic acid |
| C8H15N3O4 | |
| MFCD00133046 | |
| l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax | |
| CHEBI:73788 | |
| C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O |
Specifications
| 39537-23-0 | |
| 0.1% max | |
| Glass bottle | |
| MFCD00133046 | |
| l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax | |
| C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O | |
| 217.23 | |
| CHEBI:73788 | |
| 97% |
| 213.0°C to 220.0°C | |
| 97% | |
| C8H15N3O4 | |
| 5 g | |
| HJCMDXDYPOUFDY-WHFBIAKZSA-N | |
| (2S)-5-amino-2-[[(2S)-2-aminopropanoyl]amino]-5-oxopentanoic acid | |
| 123935 | |
| 217.22 | |
| L-Alanyl-L-glutamine |
RUO – Research Use Only