missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Alanyl-L-glutamine, 97%

Catalog No. AC451610050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC451610050 5 g Glass bottle
AC451610010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451610050 Supplier Thermo Scientific Chemicals Supplier No. 451610050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39537-23-0 | C8H15N3O4 | 217.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39537-23-0
Molecular Formula C8H15N3O4
Molecular Weight (g/mol) 217.23
MDL Number MFCD00133046
InChI Key HJCMDXDYPOUFDY-WHFBIAKZSA-N
Synonym l-alanyl-l-glutamine, ala-gln, alanyl-glutamine, alanyl glutamine, alanylglutamine, l-glutamine, l-alanyl, dipeptamin, h-ala-gln-oh, n 2-l-alanyl-l-glutamine, glutamax
PubChem CID 123935
ChEBI CHEBI:73788
IUPAC Name (2S)-5-amino-2-[[(2S)-2-aminopropanoyl]amino]-5-oxopentanoic acid
SMILES C[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 213.0°C to 220.0°C
Ignition Residue 0.1% max
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 217.22
Percent Purity 97%
Chemical Name or Material L-Alanyl-L-glutamine

RUO – Research Use Only