missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine tert-Butyl Ester Hydrochloride 97.0+%, TCI America™

Catalog No. p-7117687
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20101G 1 g
A20105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20101G Supplier TCI America Supplier No. A20101G
Only null left

Chemical Identifiers

CAS 13404-22-3
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00035524
InChI Key WIQIWPPQGWGVHD-JEDNCBNOSA-N
Synonym h-ala-otbu.hcl, tert-butyl l-alaninate hydrochloride, l-alanine tert-butyl ester hydrochloride, s-tert-butyl 2-aminopropanoate hydrochloride, tert-butyl alaninate hydrochloride, h-ala-otbu inverted exclamation mark currencyhcl, tert-butyl 2s-2-aminopropanoate hydrochloride, l-alanine t-butyl ester hydrochloride, alanine, 1,1-dimethylethyl ester, hydrochloride, l-alanine, 1,1-dimethylethyl ester, hydrochloride
PubChem CID 11275390
IUPAC Name tert-butyl (2S)-2-aminopropanoate hydrochloride
SMILES Cl.C[C@H](N)C(=O)OC(C)(C)C

Specifications

Melting Point 208°C
Color White
Quantity 1 g
Formula Weight 181.66
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanine tert-Butyl Ester Hydrochloride