missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
$750.36 - $750.36
Chemical Identifiers
| CAS | 13404-22-3 |
|---|---|
| Molecular Formula | C7H16ClNO2 |
| Molecular Weight (g/mol) | 181.66 |
| MDL Number | MFCD00035524 |
| InChI Key | WIQIWPPQGWGVHD-JEDNCBNOSA-N |
| Synonym | h-ala-otbu.hcl, tert-butyl l-alaninate hydrochloride, l-alanine tert-butyl ester hydrochloride, s-tert-butyl 2-aminopropanoate hydrochloride, tert-butyl alaninate hydrochloride, h-ala-otbu inverted exclamation mark currencyhcl, tert-butyl 2s-2-aminopropanoate hydrochloride, l-alanine t-butyl ester hydrochloride, alanine, 1,1-dimethylethyl ester, hydrochloride, l-alanine, 1,1-dimethylethyl ester, hydrochloride |
| PubChem CID | 11275390 |
| SMILES | Cl.C[C@H](N)C(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5935906
|
Thermo Scientific Chemicals
H5935906 |
5 g |
Each for $750.36
|
|
|||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Hygroscopic. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 13404-22-3 | |
| 181.66 | |
| WIQIWPPQGWGVHD-JEDNCBNOSA-N | |
| 11275390 |
| C7H16ClNO2 | |
| MFCD00035524 | |
| h-ala-otbu.hcl, tert-butyl l-alaninate hydrochloride, l-alanine tert-butyl ester hydrochloride, s-tert-butyl 2-aminopropanoate hydrochloride, tert-butyl alaninate hydrochloride, h-ala-otbu inverted exclamation mark currencyhcl, tert-butyl 2s-2-aminopropanoate hydrochloride, l-alanine t-butyl ester hydrochloride, alanine, 1,1-dimethylethyl ester, hydrochloride, l-alanine, 1,1-dimethylethyl ester, hydrochloride | |
| Cl.C[C@H](N)C(=O)OC(C)(C)C |
Specifications
| 168°C to 175°C | |
| C7H16ClNO2 | |
| 3562140 | |
| Soluble in methanol. | |
| Cl.C[C@H](N)C(=O)OC(C)(C)C | |
| 5 g | |
| 11275390 | |
| 181.66 | |
| 98% |
| 13404-22-3 | |
| MFCD00035524 | |
| h-ala-otbu.hcl, tert-butyl l-alaninate hydrochloride, l-alanine tert-butyl ester hydrochloride, s-tert-butyl 2-aminopropanoate hydrochloride, tert-butyl alaninate hydrochloride, h-ala-otbu inverted exclamation mark currencyhcl, tert-butyl 2s-2-aminopropanoate hydrochloride, l-alanine t-butyl ester hydrochloride, alanine, 1,1-dimethylethyl ester, hydrochloride, l-alanine, 1,1-dimethylethyl ester, hydrochloride | |
| WIQIWPPQGWGVHD-JEDNCBNOSA-N | |
| tert-butyl (2S)-2-aminopropanoate;hydrochloride | |
| 181.66 | |
| +1.4° (c=2 in Ethanol) | |
| Hygroscopic | |
| L-Alanine tert-butyl ester hydrochloride |
Safety and Handling
EINECSNumber : 236-497-8
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only