missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine Ethyl Ester Hydrochloride 98.0+%, TCI America™
$46.60 - $162.17
Chemical Identifiers
| CAS | 1115-59-9 |
|---|---|
| Molecular Formula | C5H12ClNO2 |
| Molecular Weight (g/mol) | 153.61 |
| MDL Number | MFCD00063662 |
| InChI Key | JCXLZWMDXJFOOI-UHFFFAOYNA-N |
| Synonym | l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride |
| PubChem CID | 2724356 |
| IUPAC Name | hydrogen ethyl 2-aminopropanoate chloride |
| SMILES | [H+].[Cl-].CCOC(=O)C(C)N |
Chemical Identifiers
| 1115-59-9 | |
| 153.61 | |
| JCXLZWMDXJFOOI-UHFFFAOYNA-N | |
| 2724356 | |
| [H+].[Cl-].CCOC(=O)C(C)N |
| C5H12ClNO2 | |
| MFCD00063662 | |
| l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride | |
| hydrogen ethyl 2-aminopropanoate chloride |
Specifications
| 1115-59-9 | |
| White | |
| MFCD00063662 | |
| l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride | |
| [H+].[Cl-].CCOC(=O)C(C)N | |
| 153.61 | |
| 153.61 | |
| Crystalline Powder |
| 79°C | |
| C5H12ClNO2 | |
| 5 g | |
| JCXLZWMDXJFOOI-UHFFFAOYNA-N | |
| hydrogen ethyl 2-aminopropanoate chloride | |
| 2724356 | |
| ≥98.0% (N) | |
| L-Alanine Ethyl Ester Hydrochloride |
Safety and Handling
TSCA : No