missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine Ethyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. A24945G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24945G 5 g
A249425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24945G Supplier TCI America Supplier No. A24945G

Chemical Identifiers

CAS 1115-59-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00063662
InChI Key JCXLZWMDXJFOOI-UHFFFAOYNA-N
Synonym l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride
PubChem CID 2724356
IUPAC Name hydrogen ethyl 2-aminopropanoate chloride
SMILES [H+].[Cl-].CCOC(=O)C(C)N

Specifications

Melting Point 79°C
Color White
Quantity 5 g
Formula Weight 153.61
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanine Ethyl Ester Hydrochloride