missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine ethyl ester hydrochloride, 98+%

CAS: 1115-59-9 | C5H12ClNO2 | 153.61 g/mol

$55.78 - $195.95

Chemical Identifiers

CAS 1115-59-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00063662
InChI Key JCXLZWMDXJFOOI-UHFFFAOYNA-N
Synonym l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride
PubChem CID 2724356
IUPAC Name ethyl (2S)-2-aminopropanoate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)C(C)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1127106
View Documents
Thermo Scientific Chemicals
A1127106
5 g
Each for $55.78
Only null left
 
AAA1127114
View Documents
Thermo Scientific Chemicals
A1127114
25 g
Each for $195.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1115-59-9
153.61
JCXLZWMDXJFOOI-UHFFFAOYNA-N
2724356
[H+].[Cl-].CCOC(=O)C(C)N
C5H12ClNO2
MFCD00063662
l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride
ethyl (2S)-2-aminopropanoate;hydrochloride

Specifications

74°C to 78°C
C5H12ClNO2
3594395
JCXLZWMDXJFOOI-UHFFFAOYNA-N
ethyl (2S)-2-aminopropanoate;hydrochloride
153.61
+2.9° (c=2.5 in water)
Hygroscopic
L-Alanine ethyl ester hydrochloride
1115-59-9
MFCD00063662
l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride
[H+].[Cl-].CCOC(=O)C(C)N
5 g
2724356
153.61
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-225-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only