missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine ethyl ester hydrochloride, 98+%

Catalog No. AAA1127114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1127114 25 g
AAA1127106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1127114 Supplier Thermo Scientific Chemicals Supplier No. A1127114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1115-59-9 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1115-59-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00063662
InChI Key JCXLZWMDXJFOOI-UHFFFAOYNA-N
Synonym l-alanine ethyl ester hydrochloride, h-ala-oet.hcl, s-ethyl 2-aminopropanoate hydrochloride, ethyl l-alaninate hydrochloride, h-ala-oet hcl, h-ala-oet hydrochloride, ethyl 2s-2-aminopropanoate hydrochloride, alanine ethyl ester hydrochloride, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride
PubChem CID 2724356
IUPAC Name ethyl (2S)-2-aminopropanoate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)C(C)N

Specifications

Melting Point 74°C to 78°C
Beilstein 3594395
Quantity 25 g
Formula Weight 153.61
Optical Rotation +2.9° (c=2.5 in water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Alanine ethyl ester hydrochloride
EINECSNumber 214-225-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only