missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 116°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 351.42 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Alanine Benzyl Ester p-Toluenesulfonate |
Safety and Handling
| TSCA | No |