missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. A20055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20055G 5 g
A200525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20055G Supplier TCI America Supplier No. A20055G

Chemical Identifiers

CAS 42854-62-6
Molecular Formula C17H21NO5S
Molecular Weight (g/mol) 351.42
MDL Number MFCD00066143,MFCD00066142
InChI Key NWOPHJSSBMABBD-UHFFFAOYNA-N
Synonym l-alanine benzyl ester 4-toluenesulfonate, l-alanine benzyl ester p-toluenesulfonate, ala-obzl tosoh, l-alanine benzyl ester p-toluenesulfonate salt, l-alanine benzyl ester tosylate, h-ala-obzl.tosoh, s-benzyl 2-aminopropanoate 4-methylbenzenesulfonate, benzyl 2s-2-aminopropanoate 4-toluenesulfonate, l-ala-obel.tsoh, benzyl 2s-2-aminopropanoate; para-toluene sulfonate
PubChem CID 2802424
IUPAC Name 4-methylbenzene-1-sulfonic acid; benzyl 2-aminopropanoate
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.CC(N)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 5 g
Formula Weight 351.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanine Benzyl Ester p-Toluenesulfonate