missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine Benzyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. A27715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27715G 5 g
A277125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27715G Supplier TCI America Supplier No. A27715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5557-83-5
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.677
MDL Number MFCD00054340
InChI Key RLMHWGDKMJIEHH-QRPNPIFTSA-N
Synonym h-ala-obzl.hcl, l-alanine benzyl ester hydrochloride, h-ala-obzl hcl, s-benzyl 2-aminopropanoate hydrochloride, l-alanine benzyl ester hcl, benzyl l-alaninate hydrochloride, benzyl 2s-2-aminopropanoate hydrochloride, h-ala-obzlhcl, ala-obzl hcl, nh2-ala-obn hcl
PubChem CID 12210893
IUPAC Name benzyl (2S)-2-aminopropanoate;hydrochloride
SMILES CC(C(=O)OCC1=CC=CC=C1)N.Cl

Specifications

Melting Point 138°C
Color White
Quantity 5 g
Formula Weight 215.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanine Benzyl Ester Hydrochloride
TSCA No
Recommended Storage Freezer