missing translation for 'onlineSavingsMsg'
Learn More

L-Alaninamide Hydrochloride 97.0+%, TCI America™

Catalog No. p-7117633
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19195G 5 g
A191925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19195G Supplier TCI America Supplier No. A19195G

Chemical Identifiers

CAS 33208-99-0
Molecular Formula C3H9ClN2O
Molecular Weight (g/mol) 124.57
MDL Number MFCD00066145
InChI Key FIAINKIUSZGVGX-DKWTVANSSA-N
Synonym l-alaninamide hydrochloride, l-alaninamide hcl, s-2-aminopropanamide hydrochloride, l-alanine amide hydrochloride, 2s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride, h-ala-nh2.hcl, ala-nh2 hcl, alaninamide, hydrochloride, l-alaninamidehydrochloride
PubChem CID 2775816
IUPAC Name (2S)-2-aminopropanamide hydrochloride
SMILES Cl.C[C@H](N)C(N)=O

Specifications

Melting Point 217°C
Color White
Quantity 5 g
Formula Weight 124.57
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-Alaninamide Hydrochloride