missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alaninamide hydrochloride, 95%
CAS: 33208-99-0 | C3H9ClN2O | 124.57 g/mol
$80.60 - $672.01
Chemical Identifiers
| CAS | 33208-99-0 |
|---|---|
| Molecular Formula | C3H9ClN2O |
| Molecular Weight (g/mol) | 124.57 |
| MDL Number | MFCD00066145 |
| InChI Key | FIAINKIUSZGVGX-DKWTVANSSA-N |
| Synonym | l-alaninamide hydrochloride, l-alaninamide hcl, s-2-aminopropanamide hydrochloride, l-alanine amide hydrochloride, 2s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride, h-ala-nh2.hcl, ala-nh2 hcl, alaninamide, hydrochloride, l-alaninamidehydrochloride |
| PubChem CID | 2775816 |
| IUPAC Name | (2S)-2-aminopropanamide;hydrochloride |
| SMILES | Cl.C[C@H](N)C(N)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6301906
|
Thermo Scientific Chemicals
H6301906 |
5 g |
Each for $80.60
|
|
|||||
|
AAH6301914
|
Thermo Scientific Chemicals
H6301914 |
25 g |
Each for $250.70
|
|
|||||
|
AAH6301922
|
Thermo Scientific Chemicals
H6301922 |
100 g |
Each for $672.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 33208-99-0 | |
| 124.57 | |
| FIAINKIUSZGVGX-DKWTVANSSA-N | |
| 2775816 | |
| Cl.C[C@H](N)C(N)=O |
| C3H9ClN2O | |
| MFCD00066145 | |
| l-alaninamide hydrochloride, l-alaninamide hcl, s-2-aminopropanamide hydrochloride, l-alanine amide hydrochloride, 2s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride, h-ala-nh2.hcl, ala-nh2 hcl, alaninamide, hydrochloride, l-alaninamidehydrochloride | |
| (2S)-2-aminopropanamide;hydrochloride |
Specifications
| 212°C to 217°C (decomposition) | |
| C3H9ClN2O | |
| l-alaninamide hydrochloride, l-alaninamide hcl, s-2-aminopropanamide hydrochloride, l-alanine amide hydrochloride, 2s-2-aminopropanamide hydrochloride, s-2-aminopropionamide hydrochloride, h-ala-nh2.hcl, ala-nh2 hcl, alaninamide, hydrochloride, l-alaninamidehydrochloride | |
| Cl.C[C@H](N)C(N)=O | |
| 5 g | |
| 2775816 | |
| 95% | |
| L-Alaninamide hydrochloride |
| 33208-99-0 | |
| MFCD00066145 | |
| FIAINKIUSZGVGX-DKWTVANSSA-N | |
| (2S)-2-aminopropanamide;hydrochloride | |
| 124.57 | |
| 124.57 | |
| Solid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only