missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Adrenaline Bitartrate 98.0+%, TCI America™
Supplier: TCI America A017525G
Specifications
| L-Adrenaline Bitartrate | |
| 154°C | |
| C13H19NO9 | |
| 25 g | |
| L-Epinephrine Bitartrate, L-Adrenaline Hydrogen Bitartrate, L-Epinephrine Hydrogen Bitartrate | |
| O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1 | |
| 333.29 | |
| 333.29 | |
| Crystalline Powder |
| 51-42-3 | |
| White-Yellow | |
| MFCD00035077 | |
| 2811 | |
| YLXIPWWIOISBDD-NDAAPVSOSA-N | |
| (2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol | |
| 124080922 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 51-42-3 | |
| 333.29 | |
| YLXIPWWIOISBDD-NDAAPVSOSA-N | |
| 124080922 | |
| O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1 |
| C13H19NO9 | |
| MFCD00035077 | |
| L-Epinephrine Bitartrate, L-Adrenaline Hydrogen Bitartrate, L-Epinephrine Hydrogen Bitartrate | |
| (2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol |
Safety and Handling
RTECSNumber : DO3500000
TSCA : No
Recommended Storage : Refrigerator