missing translation for 'onlineSavingsMsg'
Learn More

L-Adrenaline Bitartrate 98.0+%, TCI America™

Catalog No. A01755G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01755G 5 g
A01751G 1 g
A017525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A01755G Supplier TCI America Supplier No. A01755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51-42-3
Molecular Formula C13H19NO9
Molecular Weight (g/mol) 333.29
MDL Number MFCD00035077
InChI Key YLXIPWWIOISBDD-NDAAPVSOSA-N
Synonym L-Epinephrine Bitartrate, L-Adrenaline Hydrogen Bitartrate, L-Epinephrine Hydrogen Bitartrate
PubChem CID 124080922
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 333.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Adrenaline Bitartrate
RTECSNumber DO3500000
TSCA No
Recommended Storage Refrigerator