missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Adrenaline Bitartrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 154°C |
| Color | White-Yellow |
| Quantity | 1 g |
| UN Number | 2811 |
| Formula Weight | 333.29 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Adrenaline Bitartrate |
Safety and Handling
| RTECSNumber | DO3500000 |
| TSCA | No |
| Recommended Storage | Refrigerator |