missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-5-Hydroxytryptophan, 99%

C11H12N2O3

Supplier:  Thermo Scientific Chemicals 148290050

 View more versions of this product

Catalog No. AC148290050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-5-Hydroxytryptophan
Beige to White or Gray
Authentic
C11H12N2O3
MFCD00064341
15, 4886
1% max. (K.F.)
5 g
98.5 to 101.5% (on dry substance)
1ppm max.
10ppm max.
99%
Glass Bottle
-30.9° to -34.9° (20°C, 589nm) (c=1, H2O) on dry substance
0.3% max.
- 32.50 (20.00°C c=1,H2O on dry basis)
220.23
Powder

Chemical Identifiers

C11H12N2O3
MFCD00064341
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only