missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-5-Hydroxytryptophan, 99%

Catalog No. AC148290250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148290250 25 g Glass Bottle
AC148290010 1 g Glass Bottle
AC148290050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC148290250 Supplier Thermo Scientific Chemicals Supplier No. 148290250

C11H12N2O3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula C11H12N2O3
MDL Number MFCD00064341

Specifications

Arsenic (As) 1ppm max.
Color Beige to White or Gray
Heavy Metals 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Specific Rotation -30.9° to -34.9° (20°C, 589nm) (c=1, H2O) on dry substance
Merck Index 15, 4886
Sulfated Ash 0.3% max.
Water 1% max. (K.F.)
Specific Rotation Condition - 32.50 (20.00°C c=1,H2O on dry basis)
Quantity 25 g
Formula Weight 220.23
Percent Purity 98.5 to 101.5% (on dry substance)
Physical Form Powder
Chemical Name or Material L-5-Hydroxytryptophan
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only