missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-3-Phenyllactic Acid 98.0+%, TCI America™
$50.33 - $141.67
Chemical Identifiers
| CAS | 20312-36-1 |
|---|---|
| Molecular Formula | C9H9O3 |
| Molecular Weight (g/mol) | 165.17 |
| MDL Number | MFCD00004244 |
| InChI Key | VOXXWSYKYCBWHO-QMMMGPOBSA-M |
| Synonym | l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid |
| PubChem CID | 444718 |
| ChEBI | CHEBI:43065 |
| IUPAC Name | (2S)-2-hydroxy-3-phenylpropanoate |
| SMILES | O[C@@H](CC1=CC=CC=C1)C([O-])=O |
Chemical Identifiers
| 20312-36-1 | |
| 165.17 | |
| VOXXWSYKYCBWHO-QMMMGPOBSA-M | |
| 444718 | |
| (2S)-2-hydroxy-3-phenylpropanoate |
| C9H9O3 | |
| MFCD00004244 | |
| l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid | |
| CHEBI:43065 | |
| O[C@@H](CC1=CC=CC=C1)C([O-])=O |
Specifications
| 20312-36-1 | |
| White-Yellow | |
| MFCD00004244 | |
| l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid | |
| O[C@@H](CC1=CC=CC=C1)C([O-])=O | |
| 165.17 | |
| CHEBI:43065 | |
| ≥98.0% (HPLC,T) | |
| L-(-)-3-Phenyllactic Acid |
| 124°C | |
| C9H9O3 | |
| 1 g | |
| VOXXWSYKYCBWHO-QMMMGPOBSA-M | |
| (2S)-2-hydroxy-3-phenylpropanoate | |
| 444718 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No