missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-3-Phenyllactic Acid 98.0+%, TCI America™

Catalog No. P11681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11681G 1 g
P11685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11681G Supplier TCI America Supplier No. P11681G

Chemical Identifiers

CAS 20312-36-1
Molecular Formula C9H9O3
Molecular Weight (g/mol) 165.17
MDL Number MFCD00004244
InChI Key VOXXWSYKYCBWHO-QMMMGPOBSA-M
Synonym l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid
PubChem CID 444718
ChEBI CHEBI:43065
IUPAC Name (2S)-2-hydroxy-3-phenylpropanoate
SMILES O[C@@H](CC1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-3-Phenyllactic Acid