missing translation for 'onlineSavingsMsg'
Learn More

L-2-Aminoadipic Acid 97.0+%, TCI America™

Catalog No. p-7117449
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14571G 1 g
A14575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14571G Supplier TCI America Supplier No. A14571G
Only null left

Chemical Identifiers

CAS 1118-90-7
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD00002636
InChI Key OYIFNHCXNCRBQI-BYPYZUCNSA-N
Synonym l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid
PubChem CID 92136
ChEBI CHEBI:37023
IUPAC Name (2S)-2-aminohexanedioic acid
SMILES N[C@@H](CCCC(O)=O)C(O)=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 161.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-2-Aminoadipic Acid