missing translation for 'onlineSavingsMsg'
Learn More

L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, 99+%

Catalog No. p-4112162
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227800010 1 g Glass bottle
AC227800050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC227800010 Supplier Thermo Scientific Chemicals Supplier No. 227800010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 402-71-1 | C17H18ClNO3S | 351.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402-71-1
Molecular Formula C17H18ClNO3S
Molecular Weight (g/mol) 351.85
MDL Number MFCD00000935
InChI Key MQUQNUAYKLCRME-INIZCTEOSA-N
Synonym tpck, tos-phe-ch2cl, tosylphenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanine chloromethyl ketone, n-p-tosyl-l-phenylalanine chloromethyl ketone, l-1-tosylamido-2-phenylethyl chloromethyl ketone, l-n-alpha-chloroacetyl phenethyl-p-toluenesulfonamide, tos-phe-chloromethylketone, chembl60718
PubChem CID 439647
ChEBI CHEBI:9642
IUPAC Name N-[(2S)-4-chloro-3-oxo-1-phenylbutan-2-yl]-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CC1=CC=CC=C1)C(=O)CCl

Specifications

Melting Point 105°C to 109°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H5CH2CH(NHSO2C6H4CH3)COCH2Cl
Quantity 1 g
Specific Rotation Condition −86° (20°C )
Specific Rotation -86
Formula Weight 351.84
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material L-1-4′-Tosylamino-2-phenylethyl chloromethyl ketone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-954-6

RUO – Research Use Only