missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic Acid Sodium Salt, Monohydrate, HPLC Grade, 98%, Spectrum™ Chemical

Catalog No. 18600076
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600076 5 kg Poly Pail
18600072 5 g Amber Glass Bottle
18600073 25 g Amber Glass Bottle
18600074 100 g Amber Glass Bottle
18600075 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600076 Supplier Spectrum Chemical Mfg Cor Supplier No. P10435KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

P1043, 207605-40-1, C5H11O3SNa·H2O

Spectrum™ Chemical 1-Pentanesulfonic Acid Sodium Salt, Monohydrate, HPLC Grade

Chemical Identifiers

CAS 207605-40-1
Molecular Formula C5H11O3S
Molecular Weight (g/mol) 151.20
InChI Key RJQRCOMHVBLQIH-UHFFFAOYSA-M
IUPAC Name pentane-1-sulfonate
SMILES CCCCCS([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Poly Pail
Quantity 5 kg
Percent Purity 98%
Grade HPLC
Physical Form Powder