Learn More
Abcam Inc KU-55933, competitive ATM kinase inhibitor, 10MG
Catalog No. 502707406
Encompass
MW 395.5 Da, Purity >99%. Potent, selective and competitive ATM kinase inhibitor. IC values are 12.9 (ATM), 2000 (DNA-PK), 9300 (mTOR), 16600 (PI3K), >100000 (ATR) and >100000 nM (PI4K). Sensitizes cells to ionizing radiation and chemotherapeutics. Blocks Akt phosphorylation, induces G1 cell cycle arrest and induces apoptosis in breast and prostate cell lines.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502707406 Supplier Abcam Inc Supplier No. AB12063710MG
MW 395.5 Da, Purity >99%. Potent, selective and competitive ATM kinase inhibitor. IC values are 12.9 (ATM), 2000 (DNA-PK), 9300 (mTOR), 16600 (PI3K), >100000 (ATR) and >100000 nM (PI4K). Sensitizes cells to ionizing radiation and chemotherapeutics. Blocks Akt phosphorylation, induces G1 cell cycle arrest and induces apoptosis in breast and prostate cell lines.
The product is subject to the following: Abcam Restricted Use Statement