missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Krebs-Ringer Solution, bicarbonate-buffered

Catalog No. AAJ67591K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67591K2 1 L
AAJ67591AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67591K2 Supplier Thermo Scientific Chemicals Supplier No. J67591K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Physical Form Liquid
Quantity 1 L
Packaging Plastic Bottle

RUO – Research Use Only