missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Krebs-Ringer Solution, bicarbonate-buffered

Catalog No. AAJ67591AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67591AP 500 mL
AAJ67591K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67591AP Supplier Thermo Scientific Chemicals Supplier No. J67591AP

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Physical Form Liquid
Quantity 500 mL
Packaging Plastic Bottle

RUO – Research Use Only