Learn More
Leap Technologies Center Draining Screw Vial, Clear, 1.5mL, Kit Pack, 100 Pack
Catalog No. 50635211
Encompass
Kit Pack. CD Vial, 1.5-mL, 12x32mm, Screw, Clear, 9mm, Blue QuikCap, PTFE/Silicone, 100 Pack
Catalog No. 50635211 Supplier Leap Technologies Supplier No. XQS2006
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Kit Pack. CD Vial, 1.5-mL, 12x32mm, Screw, Clear, 9mm, Blue QuikCap, PTFE/Silicone, 100 Pack