missing translation for 'onlineSavingsMsg'
Learn More

KOVA™ Refractrol SP

Catalog No. 22749704
Change view
Click to view available options
Description:
Low Level
Normal Level
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
22749704 Low Level
22749705 Normal Level
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22749704 Supplier KOVA™ Supplier No. K84652

KOVA Refractrol SP tri-level control provides 5 levels of serum protein control solutions for use in refractometry

  • For use with temperature compensated and non-compensated refractometers
  • Recovery of the assigned values provides assurance of measurements of plasma or serum protein concentration
  • Each level of Refractrol is separately packaged in easy-to-use 2mL vials
Storage: Store at room temperature (18 - 30°C). Do not refrigerate or freeze. Crystals or debris may be observed in the stain, this is not uncommon for Sternheimer-Malbin stain. These artifacts may be safely removed by filtration. Stable per the expiration date on the label.

Specifications

Description Low Level
Quantity 12 x 2 mL