Learn More
Kova Plastics by Alltrista System Pac II
Complete with specially engineered microscopic slides, sample tubes , and custom pipettes or petters for reliable, simple and effective workflow standardization
Supplier: Kova Plastics by Alltrista 87155E
Description
- Complete urinalysis kit for up to 400 determinations
Package contains:
- Super Tubes (400): a 12mL graduated non-sterile disposable centrifuge tube manufactured of high-quality, break-resistant clear plastic and marked in 1mL increments
- Petters (400): a disposable plastic transfer pipette that features a unique lock for one-step, contamination-free decanting, retaining 1.0mL of urine after centrifugation
- Slide II (100): a slide with four optically clear chambers with or without grids for microscopic examination
Specifications
| Urine Collection Kit | |
| 100 Slide II slides (400 chambers), 400 Petters and 400 Super Tubes standardization | |
| 400 Pk. |