missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ KONTES™ White Polypropylene Closure for Scintillation Vial

Catalog No. K7452122400
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
K7452122400 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K7452122400 Supplier DWK Life Sciences Supplier No. 7452122400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KIMBLE™ KONTES™ White Polypropylene Closure for Scintillation Vial features a pulp-backed aluminum foil liner, is ideal for scintillation vials, and is suitable for use with various strong oxidizing agent mixtures.

Pulp-backed aluminum foil liner for scintillation vial

  • Closure is made from white polypropylene with a pulp-backed aluminum foil liner
  • 22-400 GPI thread finish
  • Top is suitable for marking
  • Suitable for use with various strong oxidizing agent mixtures
  • Ideal for scintillation vials

Specifications

Material (Liner) Aluminum Foil/Pulp
Color White
Material Polypropylene
For Use With (Application) Various strong oxidizing agent mixtures
Closure Material Polypropylene
Closure Type Screw
Quantity 1000/Cs.
For Use With (Equipment) Scintillation Vials
Autoclavable Not Autoclavable
Product Type Screw Cap