Learn More
DWK Life Sciences Kimble™ Kontes™ Vacuum Filtration Adapters
Catalog No. K1781102440
Change view
Click to view available options
Standard Taper Joint No.:
14/20
24/40
For Use With (Equipment):
Fits No. 2 pluro stopper
Fits No. 3 pluro stopper
Fits No. 4 pluro stopper
No. 5 and 6 Pluro Stopper
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No. For Use With (Equipment)
K1781102440 24/40 No. 5 and 6 Pluro Stopper
K1781200014 14/20 Fits No. 2 pluro stopper
K1781200024 24/40 Fits No. 3 pluro stopper
K1781200124 24/40 Fits No. 4 pluro stopper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. K1781102440 Supplier DWK Life Sciences Supplier No. 1781102440
Only null left
Use for reduced pressure filtration with plain stem Buchner funnels.
  • Top designed with a flange to accommodate a pluro stopper
  • Pluro stopper sold separately
  • Includes a lower drip tip
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Application) Use for reduced pressure filtration with plain stem buchner funnels
Fits Tubing ID (English) 0.37 in.
Fits Tubing ID (Metric) 9.39mm
For Use With (Equipment) No. 5 and 6 Pluro Stopper
Includes Hose connection
Material Borosilicate Glass
Product Type Vacuum Filtration Adapter
Standard Taper Joint No. 24/40