Learn More
DWK Life Sciences Kimble™ Kontes™ Stopcock Valve for Microscale Organic Chemistry Kit
One-way stopcock valve made of nylon body and HDPE plug
Supplier:  DWK Life Sciences 4201631500
Catalog No. K420163-1500
Only null left
Explore available promotions
Description
Description
One-way stopcock valve made of nylon body and HDPE plug
  • F-Luer to M-Luer
  • standard taper: 14/20
  • Included in Microscale Organic Chemistry kit
Specifications
Specifications
One-way Stopcock Valve
Nylon, HDPE
14/20
Female Luer to Male Luer
Product Suggestions
Product Suggestions