Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Full Length Stoppers
Stoppers are used with laboratory glassware such as flasks and separatory funnels.
Supplier:  DWK Life Sciences 8510004550
 View more versions of this product
Catalog No. K851000-4550
Only null left
Explore available promotions
Description
Description
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Specifications
Specifications
Solid Pennyhead Stopper
Glass
Flask and separatory funnel
45/50
Product Suggestions
Product Suggestions