missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Receiver for Midget Impinger

Catalog No. K7375520000
Change view
Click to view available options
Capacity (Metric):
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
K7375520000 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K7375520000 Supplier DWK Life Sciences Supplier No. 7375520000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KIMBLE™ KONTES™ Receiver for Midget Impinger is designed for use with midget impinger assemblies in air sampling and gas washing applications. Features a standard taper joint for secure, leak-resistant connections in laboratory collection systems.

  • Designed for use with Kimble™ Kontes™ Midget Impinger assemblies
  • Suitable for air sampling and gas washing laboratory applications
  • Standard taper joint provides secure, leak-resistant glassware connections
  • Manufactured by DWK Life Sciences to consistent quality standards

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1.01 oz.
Type Impinger Receiver
Standard Taper Joint 24/40
For Use With (Equipment) Midget Impinger