missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Reaction-Jacketed Beakers
The open top provides easy access to concentrated residual samples.
Supplier: DWK Life Sciences 3170000100
Description
The open top provides easy access to concentrated residual samples.
- A conical lower interior surface eliminates spin-out when used with an egg-shaped PTFE magnetic stir bar
- Jacket has serrated 3/8 in. hose connections
- Design meets ASTM Specification E960, Type I requirements
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Chemical Identifiers
| Connection: 0.37 in. Serrated Hose | |
| Laboratory Applications Requiring a Variety of Temperature-Controlled Reactions and Useful in Biological and Chemical Reactions Including Titrations, Enzyme Digestions and Synthetic Organic Reactions | |
| Beaker | |
| Open Top |
| Borosilicate Glass | |
| Conical Lower Interior Surface | |
| ASTM E960, ASTM E438, USP Type I |
Specifications
| 3.38 oz. | |
| 1.85 in. | |
| 4.7 cm | |
| 100 mL | |
| Laboratory Applications Requiring a Variety of Temperature-Controlled Reactions and Useful in Biological and Chemical Reactions Including Titrations, Enzyme Digestions and Synthetic Organic Reactions | |
| 3.34 in. | |
| 8.5 cm | |
| Beaker | |
| Open Top |
| Connection: 0.37 in. Serrated Hose | |
| 3.14 in. | |
| 8 cm | |
| Borosilicate Glass | |
| 2.87 in. | |
| 7.3 cm | |
| Conical Lower Interior Surface | |
| ASTM E960, ASTM E438, USP Type I |