missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Reaction-Jacketed Beakers

Catalog No. K3170000050
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
K3170000050 50 mL
K3170000100 100 mL
K3170000250 250 mL
K3170000500 500 mL
K3170001000 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. K3170000050 Supplier DWK Life Sciences Supplier No. 3170000050

The open top provides easy access to concentrated residual samples.

  • A conical lower interior surface eliminates spin-out when used with an egg-shaped PTFE magnetic stir bar
  • Jacket has serrated 3/8 in. hose connections
  • Design meets ASTM Specification E960, Type I requirements
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

Specifications

Capacity (English) 1.69 oz.
Description Connection: 0.37 in. Serrated Hose
Diameter (English) Inner 1.45 in.
Diameter (English) Outer 2.75 in.
Diameter (Metric) Inner 3.7 cm
Diameter (Metric) Outer 7 cm
Capacity (Metric) 50 mL
Material Borosilicate Glass
For Use With (Application) Laboratory Applications Requiring a Variety of Temperature-Controlled Reactions and Useful in Biological and Chemical Reactions Including Titrations, Enzyme Digestions and Synthetic Organic Reactions
Height (English) Inner 2.16 in.
Height (English) Outer 2.75 in.
Height (Metric) Inner 5.5 cm
Height (Metric) Outer 7 cm
Includes Conical Lower Interior Surface
Type Beaker
Certifications/Compliance ASTM E960, ASTM E438, USP Type I
Style Open Top
Show More Show Less