Learn More
DWK Life Sciences Kimble™ Kontes™ Polyethylene Micro Syringes
Catalog No. K7480190001
Change view
Click to view available options
Volume (Metric):
1 mL
Catalog No. Volume (Metric)
K7480190001 1 mL
1 options
Catalog No. K7480190001 Supplier DWK Life Sciences Supplier No. 7480190001
DWK Life Sciences Kimble™ Kontes™ Micro Syringes feature a Luer slip tip and are suitable for work in chromatography, nuclear medicine, HPLC sample preparation, and many laboratory procedures.
  • Polypropylene barrel
  • Polyethylene plunger
  • Latex free
  • More chemically resistant than rubber-tipped syringes
  • For research use only; not for use in diagnostic procedures
  • Sterile, individually wrapped needles contain no rubber, silicone oil, styrene, DNA, latex, or DEHP
Specifications
Material Polyethylene
Product Type Microsyringe
Graduations 0.01 cc
Volume (Metric) 1 mL