Learn More
DWK LIFE SCIENCES™ KIMBLE™ KONTES™ Pellet Pestle™ Cordless Motor
Catalog No. K7495400000
Change view
Click to view available options
Product Type:
Cordless Motor
Catalog No. Product Type
K7495400000 Cordless Motor
1 options
Catalog No. K7495400000 Supplier DWK LIFE SCIENCES™ Supplier No. 7495400000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The KIMBLE™ KONTES™ Pellet Pestle™ Cordless Motor produces more complete disaggregation than manual mixing. Compatible with both disposable and reusable Pellet Pestle™ Mixers and includes two AA batteries.
Produces more complete disaggregation than manual mixing
  • Use with either the disposable or reusable Pellet Pestle™ Mixers
  • Produces more complete disaggregation than manual mixing
  • Comes with two AA batteries included
Order Info
Replacement motor adapter available. (See Cat. No. K749541-0000)
Specifications
For Use With (Equipment) Pellet pestle mixer
Includes 2 AA batteries
Product Type Cordless Motor