Learn More
DWK Life Sciences Kimble™ Kontes™ PVDF Clamp for Heavy Duty Solution Bottle
Catalog No. 0342218A
Change view
Click to view available options
Material:
PVDF
Catalog No. Material
0342218A PVDF
1 options
Catalog No. 0342218A Supplier DWK Life Sciences Supplier No. 14950C12
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
PVDF Carboy Clamp for Heavy Duty Solution Bottle or Carboys, specifically designed to fit all sizes of KIMAX™ 14950 solution carboy bottles.
  • Non-sterile, autoclavable PVDF clamp
Specifications
Product Type Threaded Clamp
Material PVDF
For Use With (Equipment) Heavy duty solution bottle