Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Reusable Threaded Centrifuge Tubes
Heavy wall withstands high centrifuge speeds
Supplier:  DWK Life Sciences 4521250
 View more versions of this product
Catalog No. 0555812B
Only null left
Description
Description
Heavy wall withstands high centrifuge speeds
Ideal for separating neutralized sediment for culturing, from specimens of sputum in clinical work and microbiological applications.
  • Marking spot is sandblasted
  • Caps are not attached
  • Supplied with autoclavable caps with rubber liners
  • GPI thread finish is 24mm-410
Specifications
Chemical Identifiers
USP, ASTM E 438
Useful in separating neutralized sediment for culturing from specimens of sputum in clinical work and microbiological applications
24-410
Round
Marking Spot
Centrifuge Tube
Borosilicate Glass
Rubber
Autoclavable
Round
29 mm
3550 x g
Specifications
USP, ASTM E 438
Borosilicate Glass
Rubber
Autoclavable
123 mm
Round
29 mm
3550 x g
50 mL
Useful in separating neutralized sediment for culturing from specimens of sputum in clinical work and microbiological applications
24-410
Round
1.69 oz.
Marking Spot
Centrifuge Tube
Product Suggestions
Product Suggestions