Learn More
DWK Life Sciences Kimble™ Kontes™ Hose Inlet Adapter with Fritted Disc
Ideal for use as a vent or for the introduction of pressure/vacuum.
Supplier: DWK Life Sciences 1829202440
Description
- Adapter has a 170-220 micron porosity disc between the hose connection and the inner joint
- Fritted disc prevents solids from being introduced into the system
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
| USP Type I, ASTM E438, Type I, Class A | |
| 9.39mm | |
| 95mm | |
| Borosilicate Glass | |
| 24/40 |
| 0.37 in. | |
| 3.74 in. | |
| Fritted discs | |
| Hose Inlet Adapter |