Learn More
DWK Life Sciences Kimble™ Kontes™ HPLC Ultra-Ware™ Standard Filtration/Delivery Four-Valve Cap System
Catalog No. K9539810005
Change view
Click to view available options
Product Type:
Check Valve
Screw Cap
Stem Valve
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
K9539810005 Screw Cap
K9539810001 Stem Valve
K9539810003 Check Valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K9539810005 Supplier DWK Life Sciences Supplier No. 9539810005
Parts and accessories for standard four-valve cap system
  • Lower cap retainer ring
  • Valve stem, CTFE, short (for sparge and pump ports)
  • Check valve with retainer
  • Lower screw cap, GL 45
Specifications
No. per Case 1 Cs.
Thread Size (Metric) GL 45
Product Type Screw Cap
For Use With (Equipment) Ultra-ware™ standard filtration/delivery four valve cap system