Learn More
DWK Life Sciences Kimble™ Kontes™ Four-Neck Round-Bottom Heavy-Wall Flasks with Vertical Necks
With standard taper outer joints and straight side necks
Supplier: DWK Life Sciences 6070001529
Description
- Heavy wall
- Four neck round-bottom flask
- Outer joints all vertical
Specifications
| 3000 mL | |
| Vertical | |
| 29/42 |
| Borosilicate Glass | |
| 4/Cs. | |
| 185 mm |