Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column ETFE Nut for 1/16 in. Tubing
Catalog No. K4208210116
Change view
Click to view available options
For Use With:
Flex-Column
Catalog No. For Use With
K4208210116 Flex-Column
1 options
Catalog No. K4208210116 Supplier DWK Life Sciences Supplier No. 4208210116
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™ and DISPOSAFLEX™ columns.
  • ETFE Flangeless Nut
  • 1/4-28 Thread, for 1/16 in. Tubing
  • Use with the 420822-Series Flangeless Ferrules
  • Case of 1
Specifications
Type Flangeless Nut
Diameter (English) Outer 0.0625 in.
Diameter (Metric) Outer 0.158 cm
Thread Size 0.25-28
For Use With Flex-Column
Material ETFE