Learn More
DWK Life Sciences Kimble™ Flex-Column Chromatography Fittings: Adapter with 1/4-28 MaleThread to Male Luer
Catalog No. K4204072113
Change view
Click to view available options
For Use With:
Flex-Column
Catalog No. For Use With
K4204072113 Flex-Column
1 options
Catalog No. K4204072113 Supplier DWK Life Sciences Supplier No. 4204072113
This ETFE FLEX-COLUMN™ fitting is a male adapter with 1/4-28 thread to male Luer.
  • ETFE FLEX-COLUMN™ Fitting
  • Male Luer to Male 1/4 in.-28 Thread
  • Case of 1
Specifications
Type Adapter
Thread Size 0.25-28
Port Type Male luer to male
For Use With Flex-Column
Material ETFE