Learn More
DWK Life Sciences Kimble™ Flex-Column Chromatography Fittings: Adapter with 1/4-28 Female Thread to Female Luer
Catalog No. K4204072003
Change view
Click to view available options
For Use With:
Flex-Column
Catalog No. For Use With
K4204072003 Flex-Column
1 options
Catalog No. K4204072003 Supplier DWK Life Sciences Supplier No. 4204072003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This ETFE FLEX-COLUMN™ fitting is a female adapter with 1/4-28 thread to female Luer.
  • ETFE FLEX-COLUMN™ Fitting
  • Female Luer to Female 1/4 in.-28 Thread
  • Case of 1
Specifications
Type Adapter
Thread Size 0.25-28
Port Type Female luer to female
For Use With Flex-Column
Material ETFE