Learn More
DWK Life Sciences Kimble™ Kontes™ Distillation Flasks
Improved version of classic Schlenk tube
Supplier: DWK Life Sciences 2131001014
Description
- Modified Kjeldahl shape facilitates heating, stirring, and solvent stripping
- Easily transfer liquids and solids
- Glass plug
- Pressure/vacuum retainer
Specifications
| 100 mL | |
| For heating, stirring and solvent stripping without impairing ease of liquid and solid transfer | |
| 15.5 cm | |
| Kjeldahl | |
| Round |
| Borosilicate Glass | |
| 6.1 in. | |
| Glass plug and the patented pressure/vacuum retainer | |
| 14/20 |