Learn More
DWK Life Sciences Kimble™ Kontes™ Disposaflex™ Lower Column Fitting
Description
- Includes recess to hold polyethylene bed support or glass wool plug
- Polypropylene
- Pack of 100
Specifications
Specifications
| Material | Polypropylene |