Learn More
DWK Life Sciences Kimble™ Kontes™ Arsine Generator Absorber Tube
Catalog No. K2411020000
Change view
Click to view available options
Standard Taper Joint:
12/2
Catalog No. Standard Taper Joint
K2411020000 12/2
1 options
Catalog No. K2411020000 Supplier DWK Life Sciences Supplier No. 2411020000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This Arsine Generator Absorber Tube is a Component Part For Arsine Generators
  • Arsine Generator Absorber Tube Only
  • Spherical Joint 12/2
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
Specifications
Product Type Absorber Tube
Standard Taper Joint 12/2
For Use With (Equipment) For arsine generator